Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanedioic-2,2,7,7-d4 acid | 19031-57-3 | 99.9% | 178.22 g/mol | C8H10D4O4 | 100 MG
SDP

Catalog No. 5000302623
Encompass_Preferred

Suberic acid-d4 is the deuterium-labeled analogue of suberic (octanedioic) acid, provided as a high-purity isotopic standard for analytical and metabolic studies. It is intended for use as an internal standard in quantitative mass spectrometry, tracer experiments, and analytical method development.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High chemical purity (~99.9%) and isotopic enrichment.
  • Molecular weight 178.22 g/mol and formula C8H10D4O4.
  • Soluble in DMSO at 100 mg/mL for easy stock preparation.
  • Stable as powder at -20°C for long-term storage.
  • Available in small mg-scale quantities suitable for analytical workflows.

Catalog No. 50-003-02623 Supplier Medchemexpress LLC Supplier No. HYW015300S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suberic acid-d4 is the deuterium-labeled analogue of suberic (octanedioic) acid, provided as a high-purity isotopic standard for analytical and metabolic studies. It is intended for use as an internal standard in quantitative mass spectrometry, tracer experiments, and analytical method development.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High chemical purity (~99.9%) and isotopic enrichment.
  • Molecular weight 178.22 g/mol and formula C8H10D4O4.
  • Soluble in DMSO at 100 mg/mL for easy stock preparation.
  • Stable as powder at -20°C for long-term storage.
  • Available in small mg-scale quantities suitable for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.