Learn More
Medchemexpress LLC Octanedioic acid, 1,8-bis(2,5-dioxo-3-sulfo-1-pyrrolidinyl) ester | 82436-77-9 | 98.0% | 528.47 g/mol | C16H20N2O14S2 | 10 MG
SDP
Catalog No. 5000202974
Encompass_Preferred
BS3 Crosslinker is a water-soluble, non-cleavable, amine-reactive homobifunctional crosslinker (sulfo-NHS ester) designed for bioconjugation and protein crosslinking in aqueous systems. It selectively reacts with primary amines to form stable amide bonds and is commonly used for protein structural studies, surface crosslinking, and as a linker in antibody-drug conjugate research.
  • Water-soluble sulfo-NHS ester for use in aqueous buffers.
  • Reacts with primary amines to form stable amide bonds.
  • Non-cleavable linker for stable, long-lived conjugates.
  • Membrane-impermeable, suitable for surface and extracellular crosslinking.
  • Useful for bioconjugation, protein structural analysis, and linker development.
Catalog No. 50-002-02974 Supplier Medchemexpress LLC Supplier No. HY12432910MG
BS3 Crosslinker is a water-soluble, non-cleavable, amine-reactive homobifunctional crosslinker (sulfo-NHS ester) designed for bioconjugation and protein crosslinking in aqueous systems. It selectively reacts with primary amines to form stable amide bonds and is commonly used for protein structural studies, surface crosslinking, and as a linker in antibody-drug conjugate research.
  • Water-soluble sulfo-NHS ester for use in aqueous buffers.
  • Reacts with primary amines to form stable amide bonds.
  • Non-cleavable linker for stable, long-lived conjugates.
  • Membrane-impermeable, suitable for surface and extracellular crosslinking.
  • Useful for bioconjugation, protein structural analysis, and linker development.