Learn More
Medchemexpress LLC Octanedioic acid, 1,8-bis(2,5-dioxo-3-sulfo-1-pyrrolidinyl) ester | 82436-77-9 | 98.0% | 528.47 g·mol⁻¹ | C16H20N2O14S2 | 50 MG
SDP
Catalog No. 5000204283
Encompass_Preferred
BS3 Crosslinker is a water-soluble, non-cleavable homobifunctional crosslinker that reacts with primary amines to form stable amide bonds. It contains sulfo-NHS ester groups that enable efficient conjugation in aqueous buffers, making it suitable for protein crosslinking and antibody-drug conjugate synthesis in research applications.
  • Water soluble and membrane impermeable, enabling reactions in aqueous buffers.
  • Reacts with primary amines via NHS-ester chemistry for efficient conjugation.
  • Forms stable, non-cleavable amide bonds for durable linkages.
  • Compatible with antibody-drug conjugate (ADC) and protein crosslinking workflows.
  • Supplied with defined purity and characterized molecular weight for reproducibility.
  • Available in small lab-friendly pack sizes for research use.
Catalog No. 50-002-04283 Supplier Medchemexpress LLC Supplier No. HY12432950MG
BS3 Crosslinker is a water-soluble, non-cleavable homobifunctional crosslinker that reacts with primary amines to form stable amide bonds. It contains sulfo-NHS ester groups that enable efficient conjugation in aqueous buffers, making it suitable for protein crosslinking and antibody-drug conjugate synthesis in research applications.
  • Water soluble and membrane impermeable, enabling reactions in aqueous buffers.
  • Reacts with primary amines via NHS-ester chemistry for efficient conjugation.
  • Forms stable, non-cleavable amide bonds for durable linkages.
  • Compatible with antibody-drug conjugate (ADC) and protein crosslinking workflows.
  • Supplied with defined purity and characterized molecular weight for reproducibility.
  • Available in small lab-friendly pack sizes for research use.