Learn More
Medchemexpress LLC Octanedioic acid, 1,8-bis(2,5-dioxo-3-sulfo-1-pyrrolidinyl) ester | 82436-77-9 | 99.8% | 528.47 g/mol | C16H20N2O14S2 | 25 MG
SDP
Catalog No. 5000203752
Encompass_Preferred
BS3 Crosslinker is a water-soluble, non-cleavable, homobifunctional amine crosslinker that reacts with primary amines to form stable amide bonds. It is intended for research use in antibody-drug conjugation, protein crosslinking, and bioconjugation workflows, and is supplied as a high-purity solid with recommended cold storage for long-term stability.
  • Water-soluble, homobifunctional amine crosslinker.
  • Non-cleavable linker for stable conjugates.
  • Forms stable amide bonds with primary amines.
  • High purity suitable for research applications.
  • Available in small pack sizes for laboratory use.
  • Store sealed at low temperature to preserve stability.
Catalog No. 50-002-03752 Supplier Medchemexpress LLC Supplier No. HY12432925MG
BS3 Crosslinker is a water-soluble, non-cleavable, homobifunctional amine crosslinker that reacts with primary amines to form stable amide bonds. It is intended for research use in antibody-drug conjugation, protein crosslinking, and bioconjugation workflows, and is supplied as a high-purity solid with recommended cold storage for long-term stability.
  • Water-soluble, homobifunctional amine crosslinker.
  • Non-cleavable linker for stable conjugates.
  • Forms stable amide bonds with primary amines.
  • High purity suitable for research applications.
  • Available in small pack sizes for laboratory use.
  • Store sealed at low temperature to preserve stability.