Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanedioic acid,1-methyl ester | 3946-32-5 | 188.22 | 1 ML
SDP

Catalog No. 5000448874
Encompass_Preferred

Monomethyl octanoate (also known as Suberic acid monomethyl ester) is an ester product primarily utilized as a PROTAC linker. It can be employed in the synthesis of PROTACs, specifically DDD2 (HY-176261), for research purposes.

  • Ester product
  • Functions as a PROTAC linker
  • Suitable for the synthesis of PROTACs, including DDD2
  • High purity for reliable research
  • Molecular formula: C9H16O4
  • Molecular weight: 188.22
  • Appears as colorless to light yellow
  • Solid below 17°C, liquid above 19°C
  • Classified as ketones, aldehydes, and acids
  • Derived from an endogenous metabolite

Catalog No. 50-004-48874 Supplier Medchemexpress LLC Supplier No. HYW00195810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl octanoate (also known as Suberic acid monomethyl ester) is an ester product primarily utilized as a PROTAC linker. It can be employed in the synthesis of PROTACs, specifically DDD2 (HY-176261), for research purposes.

  • Ester product
  • Functions as a PROTAC linker
  • Suitable for the synthesis of PROTACs, including DDD2
  • High purity for reliable research
  • Molecular formula: C9H16O4
  • Molecular weight: 188.22
  • Appears as colorless to light yellow
  • Solid below 17°C, liquid above 19°C
  • Classified as ketones, aldehydes, and acids
  • Derived from an endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.