Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanedioic acid-d12 | 169397-99-3 | 98.6% | 186.27 | C8H2D12O4 | 5 MG
SDP

Catalog No. 5000303224
Encompass_Preferred

Suberic acid-d12 (octanedioic acid-d12) is the deuterium-labeled form of suberic acid (CAS 169397-99-3) supplied as a white to off-white solid for laboratory research. It is intended for use as an isotopically labeled standard or tracer in analytical, metabolic, and mass spectrometry applications and is provided with a certificate of analysis confirming purity and characterization.

  • Deuterium-labeled octanedioic acid suitable for tracer and internal standard use.
  • CAS 169397-99-3.
  • Chemical formula C8H2D12O4; molecular weight 186.27.
  • Purity 98.6% as supplied.
  • White to off-white solid appearance.
  • Supplied in small research pack sizes suitable for analytical applications.
  • Intended for research use only, not for human or clinical use.

Catalog No. 50-003-03224 Supplier Medchemexpress LLC Supplier No. HYW015300S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suberic acid-d12 (octanedioic acid-d12) is the deuterium-labeled form of suberic acid (CAS 169397-99-3) supplied as a white to off-white solid for laboratory research. It is intended for use as an isotopically labeled standard or tracer in analytical, metabolic, and mass spectrometry applications and is provided with a certificate of analysis confirming purity and characterization.

  • Deuterium-labeled octanedioic acid suitable for tracer and internal standard use.
  • CAS 169397-99-3.
  • Chemical formula C8H2D12O4; molecular weight 186.27.
  • Purity 98.6% as supplied.
  • White to off-white solid appearance.
  • Supplied in small research pack sizes suitable for analytical applications.
  • Intended for research use only, not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.