Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octanohydroxamic Acid 99.0+%, TCI America™
SDP

Catalog No. C123225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C123225G 25 g
C12325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C123225G Supplier TCI America Supplier No. C123225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7377-03-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00143918
InChI Key RGUVUPQQFXCJFC-UHFFFAOYSA-N
Synonym Caprylohydroxamic Acid, N-Hydroxyoctanamide
PubChem CID 23846
IUPAC Name N-hydroxyoctanamide
SMILES CCCCCCCC(=O)NO

Specifications

Melting Point 78°C
Color White
Quantity 25 g
Formula Weight 159.23
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Octanohydroxamic Acid
RTECSNumber RG9900500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.