Learn More
Medchemexpress LLC Octanoic acid | 124-07-2 | 99.0% | 144.21 | 100 MG
SDP
Catalog No. 5000404551
Encompass_Preferred
Octanoic acid (Standard) is the analytical standard of Octanoic acid, intended for research and analytical applications. It is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes.
  • Analytical and reference standard
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Oily liquid with a slightly unpleasant rancid taste
  • Commercial use in perfumery ester production
  • Commercial use in dye manufacturing
Catalog No. 50-004-04551 Supplier Medchemexpress LLC Supplier No. HY41417R100MG
Octanoic acid (Standard) is the analytical standard of Octanoic acid, intended for research and analytical applications. It is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes.
  • Analytical and reference standard
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Oily liquid with a slightly unpleasant rancid taste
  • Commercial use in perfumery ester production
  • Commercial use in dye manufacturing