Learn More
Medchemexpress LLC Octanoic acid, 2-(2-propyn-1-yl)- | 96017-59-3 | MFCD00946863 | 98.0% | 182.26 g/mol | C11H18O2 | 10 MG
SDP
Catalog No. 5000230090
Encompass_Preferred
2-Hexyl-4-pentynoic acid is an alkynyl-substituted octanoic acid used in biochemical and cell-based research as a histone deacetylase (HDAC) inhibitor. The compound contains a terminal alkyne for click-chemistry applications and is supplied as a colorless to light yellow liquid at high reported purity.
  • Alkynyl-substituted octanoic acid with HDAC inhibitory activity.
  • Terminal alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High reported purity (98.0%) and supplied as a liquid.
  • Small packaging options available, including 10 mg quantities for research use.
  • CAS number 96017-59-3; molecular weight 182.26 g/mol.
Catalog No. 50-002-30090 Supplier Medchemexpress LLC Supplier No. HY11878310MG
2-Hexyl-4-pentynoic acid is an alkynyl-substituted octanoic acid used in biochemical and cell-based research as a histone deacetylase (HDAC) inhibitor. The compound contains a terminal alkyne for click-chemistry applications and is supplied as a colorless to light yellow liquid at high reported purity.
  • Alkynyl-substituted octanoic acid with HDAC inhibitory activity.
  • Terminal alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High reported purity (98.0%) and supplied as a liquid.
  • Small packaging options available, including 10 mg quantities for research use.
  • CAS number 96017-59-3; molecular weight 182.26 g/mol.