Learn More
Medchemexpress LLC Octanoic acid, 2-butyl- | 27610-92-0 | MFCD00661074 | 100.0% | 200.32 g/mol | C12H24O2 | 25 G
SDP
Catalog No. 5000303449
Encompass_Preferred
2-Butyloctanoic acid is an organic biochemical reagent (CAS 27610-92-0) used in life science research as a building block or analytical reference. It is available as a neat compound and as a solution in DMSO, and is supplied with documentation intended for research use only.
  • High purity suitable for analytical and research applications.
  • Supplied as a solid and as a 10 mM solution in DMSO.
  • Provided with certificate of analysis and safety data sheet.
  • Stable at room temperature as a solid; store solutions at -20°C or -80°C for long-term storage.
  • Molecular formula C12H24O2 and molecular weight 200.32 g/mol.
Catalog No. 50-003-03449 Supplier Medchemexpress LLC Supplier No. 25GHYW014887
Add to Cart
Add to Cart
2-Butyloctanoic acid is an organic biochemical reagent (CAS 27610-92-0) used in life science research as a building block or analytical reference. It is available as a neat compound and as a solution in DMSO, and is supplied with documentation intended for research use only.
  • High purity suitable for analytical and research applications.
  • Supplied as a solid and as a 10 mM solution in DMSO.
  • Provided with certificate of analysis and safety data sheet.
  • Stable at room temperature as a solid; store solutions at -20°C or -80°C for long-term storage.
  • Molecular formula C12H24O2 and molecular weight 200.32 g/mol.