Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoic acid, 2-hydroxy- | 617-73-2 | MFCD00014410 | 98.0% | 160.21 g·mol⁻¹ | C8H16O3 | 50 MG
SDP

Catalog No. 5000303743
Encompass_Preferred

2-Hydroxyoctanoic acid is a hydroxy fatty acid used in biochemical research as an inhibitor of medium-chain acyl-CoA synthetase (reported Ki ≈ 500 μM). It is supplied as a solid and as a DMSO solution, with supporting documentation for characterization and safe handling. High purity and available analytical data make it suitable for metabolism and enzymology studies.

  • Inhibits medium-chain acyl-CoA synthetase (Ki ≈ 500 μM).
  • Available as solid and 10 mM solution in DMSO for convenient use.
  • Purity 98.0% suitable for research applications.
  • Molecular weight 160.21 g·mol⁻¹ and formula C8H16O3.
  • Documentation includes datasheet, COA, and SDS.

Catalog No. 50-003-03743 Supplier Medchemexpress LLC Supplier No. HYW01544450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxyoctanoic acid is a hydroxy fatty acid used in biochemical research as an inhibitor of medium-chain acyl-CoA synthetase (reported Ki ≈ 500 μM). It is supplied as a solid and as a DMSO solution, with supporting documentation for characterization and safe handling. High purity and available analytical data make it suitable for metabolism and enzymology studies.

  • Inhibits medium-chain acyl-CoA synthetase (Ki ≈ 500 μM).
  • Available as solid and 10 mM solution in DMSO for convenient use.
  • Purity 98.0% suitable for research applications.
  • Molecular weight 160.21 g·mol⁻¹ and formula C8H16O3.
  • Documentation includes datasheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.