Learn More
Medchemexpress LLC HY-41417 500mg Medchemexpress, Octanoic acid CAS:124-07-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY41417500MG
Encompass_Preferred
Medchemexpress, HY-41417 500mg Octanoic acid CAS:124-07-2 Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and used commercially in the production of esters used in perfumery and also in the manufacture of dyes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2238