Learn More
Medchemexpress LLC Octanoic acid, 8-[(1,1-dimethylethoxy)carbonyl]amino | 30100-16-4 | MFCD00270350 | ≥97.0% | 259.34 g/mol | C13H25NO4 | 25 G
SDP
Catalog No. 5000209751
Encompass_Preferred
N-Boc-8-amino-octanoic acid is a Boc-protected amino acid with an eight-carbon aliphatic spacer and a terminal carboxylic acid. It is used as a flexible linker in PROTAC and peptide synthesis; the carboxylic acid can be coupled to amines to form amide bonds, and the Boc protecting group can be removed under mild acidic conditions to reveal the free amine.
  • Boc-protected amino acid suitable for peptide and PROTAC linker synthesis.
  • Eight-carbon aliphatic spacer provides flexible linkage.
  • Terminal carboxylic acid reactive for amide bond formation.
  • Boc group removable under mild acidic conditions.
  • Purity ≥97.0% and white to off-white solid appearance.
  • Available in multiple package sizes, including 25 G.
Catalog No. 50-002-09751 Supplier Medchemexpress LLC Supplier No. 25GHYW041856
N-Boc-8-amino-octanoic acid is a Boc-protected amino acid with an eight-carbon aliphatic spacer and a terminal carboxylic acid. It is used as a flexible linker in PROTAC and peptide synthesis; the carboxylic acid can be coupled to amines to form amide bonds, and the Boc protecting group can be removed under mild acidic conditions to reveal the free amine.
  • Boc-protected amino acid suitable for peptide and PROTAC linker synthesis.
  • Eight-carbon aliphatic spacer provides flexible linkage.
  • Terminal carboxylic acid reactive for amide bond formation.
  • Boc group removable under mild acidic conditions.
  • Purity ≥97.0% and white to off-white solid appearance.
  • Available in multiple package sizes, including 25 G.