Learn More
Medchemexpress LLC Octanoic acid, 8-amino- | 1002-57-9 | MFCD00008245 | ≥98.0% | 159.23 g/mol | C8H17NO2 | 50 G
SDP
Catalog No. 5000301299
Encompass_Preferred
8-Aminooctanoic acid is an omega-amino fatty acid (octanoic acid bearing an amino group at the ω-position) supplied as a research reagent and analytical standard. It is commonly used in biochemical studies, peptide modification, and as an analytical reference; product listings indicate purity ≥98.0% and available COA documentation.
  • Molecular formula C8H17NO2 and molecular weight 159.23 g/mol.
  • High purity (≥98.0%) suitable for research applications.
  • Functions as an ω-amino fatty acid for biochemical and metabolic studies.
  • Useful as a structural modification tool in peptide synthesis to restrict conformation.
  • Supplied with datasheet and certificate of analysis for quality verification.
Catalog No. 50-003-01299 Supplier Medchemexpress LLC Supplier No. HYW01867850G
Add to Cart
Add to Cart
8-Aminooctanoic acid is an omega-amino fatty acid (octanoic acid bearing an amino group at the ω-position) supplied as a research reagent and analytical standard. It is commonly used in biochemical studies, peptide modification, and as an analytical reference; product listings indicate purity ≥98.0% and available COA documentation.
  • Molecular formula C8H17NO2 and molecular weight 159.23 g/mol.
  • High purity (≥98.0%) suitable for research applications.
  • Functions as an ω-amino fatty acid for biochemical and metabolic studies.
  • Useful as a structural modification tool in peptide synthesis to restrict conformation.
  • Supplied with datasheet and certificate of analysis for quality verification.