Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoic acid, 8-bromo-, 1-octylnonyl ester | 2089253-22-3 | MFCD34178802 | 97.0% | 461.56 | C25H49BrO2 | 100 MG
SDP

Catalog No. 5000281458
Encompass_Preferred

Heptadecan-9-yl 8-bromooctanoate is a brominated lipid ester intended for research use as a building block or modifier in lipid nanoparticle and liposome formulations. As an ester of 8-bromooctanoic acid with a long alkyl alcohol, it combines a hydrophobic tail with a reactive brominated site useful for formulation or synthetic modification.

  • Suitable for incorporation into lipid nanoparticles and liposomes.
  • Provides a long hydrophobic alkyl chain for membrane integration.
  • Contains a brominated functional group for covalent modification.
  • Supplied with batch-specific purity to support reproducible results.
  • Soluble in common organic solvents for formulation and synthesis workflows.

Catalog No. 50-002-81458 Supplier Medchemexpress LLC Supplier No. HYW440813100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heptadecan-9-yl 8-bromooctanoate is a brominated lipid ester intended for research use as a building block or modifier in lipid nanoparticle and liposome formulations. As an ester of 8-bromooctanoic acid with a long alkyl alcohol, it combines a hydrophobic tail with a reactive brominated site useful for formulation or synthetic modification.

  • Suitable for incorporation into lipid nanoparticles and liposomes.
  • Provides a long hydrophobic alkyl chain for membrane integration.
  • Contains a brominated functional group for covalent modification.
  • Supplied with batch-specific purity to support reproducible results.
  • Soluble in common organic solvents for formulation and synthesis workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.