Learn More
Medchemexpress LLC Octanoic acid, 8-bromo-, 1-octylnonyl ester | 2089253-22-3 | MFCD34178802 | 97.0% | 461.56 g/mol | C25H49BrO2 | 25 MG
SDP
Catalog No. 5000281680
Encompass_Preferred
Heptadecan-9-yl 8-bromooctanoate is an alkyl bromooctanoate ester used as a reagent for construction and modification of lipid nanoparticles and related lipid chemistries in research settings. It is supplied as a small-scale laboratory reagent and is characterized by a certificate of analysis reporting identity and purity suitable for formulation work.
  • Useful for building or modifying lipid nanoparticles.
  • Suitable for lipid chemistry and formulation research.
  • Supplied in small laboratory-scale quantities for experimental use.
  • High purity: 97.0% (NMR) as reported on COA.
  • Chemical formula C25H49BrO2; molecular weight 461.56 g/mol.
Catalog No. 50-002-81680 Supplier Medchemexpress LLC Supplier No. HYW44081325MG
Heptadecan-9-yl 8-bromooctanoate is an alkyl bromooctanoate ester used as a reagent for construction and modification of lipid nanoparticles and related lipid chemistries in research settings. It is supplied as a small-scale laboratory reagent and is characterized by a certificate of analysis reporting identity and purity suitable for formulation work.
  • Useful for building or modifying lipid nanoparticles.
  • Suitable for lipid chemistry and formulation research.
  • Supplied in small laboratory-scale quantities for experimental use.
  • High purity: 97.0% (NMR) as reported on COA.
  • Chemical formula C25H49BrO2; molecular weight 461.56 g/mol.