Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoic acid, 8-bromo-, 1-octylnonyl ester | 2089253-22-3 | MFCD34178802 | 97.0% | 461.56 g/mol | C25H49BrO2 | 50 MG
SDP

Catalog No. 5000281166
Encompass_Preferred

Heptadecan-9-yl 8-bromooctanoate is a brominated lipid ester supplied for research use as a hydrophobic building block for lipid nanoparticles (LNPs) and related lipid formulations. It is used in lipid chemistry and formulation development to introduce a brominated alkyl chain for downstream modification and formulation studies.

  • Brominated lipid ester for LNP construction.
  • Useful as a hydrophobic building block in lipid formulations.
  • Chemical formula C25H49BrO2; molecular weight 461.56 g/mol.
  • CAS number 2089253-22-3 for unambiguous identification.
  • Purity 97.0% as supplied.
  • Storage: pure form -20°C up to 3 years; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes including solution and solid forms.

Catalog No. 50-002-81166 Supplier Medchemexpress LLC Supplier No. HYW44081350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heptadecan-9-yl 8-bromooctanoate is a brominated lipid ester supplied for research use as a hydrophobic building block for lipid nanoparticles (LNPs) and related lipid formulations. It is used in lipid chemistry and formulation development to introduce a brominated alkyl chain for downstream modification and formulation studies.

  • Brominated lipid ester for LNP construction.
  • Useful as a hydrophobic building block in lipid formulations.
  • Chemical formula C25H49BrO2; molecular weight 461.56 g/mol.
  • CAS number 2089253-22-3 for unambiguous identification.
  • Purity 97.0% as supplied.
  • Storage: pure form -20°C up to 3 years; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes including solution and solid forms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.