Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018957 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8957 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8957 Supplier MilliporeSigma Supelco Supplier No. 216395ML
Add to Cart
Add to Cart

Octanoic acid is a medium chain saturated fatty acid (MCFA). It is a nutrient present in dairy products and specific oils like palm kernel and coconut oils.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 124-07-2
Vapor Pressure 1 mmHg (78°C)
Vapor Density 5 (vs. air)
Density 0.91 g/mL (at 25°C)
Flash Point >110°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)6 COOH
MDL Number MFCD00004429
Melting Point 15°C to 17°C
Boiling Point 237°C; 237°C to 238°C
Synonym Acid C8; Caprylic acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 144.21
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.