Learn More
Medchemexpress LLC Octanoic acid (caprylic acid) | 124-07-2 | 99.85% | 144.21 | 10 G
SDP
Catalog No. 5000404933
Encompass_Preferred
Octanoic acid, also known as Caprylic acid, is an oily liquid utilized in commercial applications such as the production of esters for perfumery and dyes. It is classified as a human endogenous metabolite and has demonstrated various biological activities, including acting as a tremor-suppressing agent.
  • Used in commercial production of esters for perfumery and dyes.
  • Functions as a tremor-suppressing agent.
  • Potentiates glucose-stimulated insulin secretion in MIN6 cells.
  • Induces neurite outgrowth in PC12 cells.
  • Inhibits harmaline tremor in mouse models.
  • Shows protective effects in Parkinson's disease models.
  • Promotes branched-chain amino acid catabolism.
Catalog No. 50-004-04933 Supplier Medchemexpress LLC Supplier No. HY4141710G
Octanoic acid, also known as Caprylic acid, is an oily liquid utilized in commercial applications such as the production of esters for perfumery and dyes. It is classified as a human endogenous metabolite and has demonstrated various biological activities, including acting as a tremor-suppressing agent.
  • Used in commercial production of esters for perfumery and dyes.
  • Functions as a tremor-suppressing agent.
  • Potentiates glucose-stimulated insulin secretion in MIN6 cells.
  • Induces neurite outgrowth in PC12 cells.
  • Inhibits harmaline tremor in mouse models.
  • Shows protective effects in Parkinson's disease models.
  • Promotes branched-chain amino acid catabolism.