Learn More
Medchemexpress LLC Octanoic acid (Caprylic acid) | 124-07-2 | 99.9% | 144.21 | 1 ML
SDP
Catalog No. 5000404425
Encompass_Preferred
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. It also acts as a tremor-suppressing agent.
  • Used in perfumery and dye manufacturing
  • Functions as a tremor-suppressing agent
  • Potentiates glucose-stimulated insulin secretion (GSIS) in MIN6 cells
  • Induces neurite outgrowth in PC12 cells and activates p38 MAPK and ERK pathways
  • Inhibits harmaline tremor in a mouse model of essential tremor
  • Shows protective effects in the mouse model of PD induced by MPTP
  • Promotes branched-chain amino acid (BCAA) catabolism in rats
Catalog No. 50-004-04425 Supplier Medchemexpress LLC Supplier No. HY4141710MM/1ML
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. It also acts as a tremor-suppressing agent.
  • Used in perfumery and dye manufacturing
  • Functions as a tremor-suppressing agent
  • Potentiates glucose-stimulated insulin secretion (GSIS) in MIN6 cells
  • Induces neurite outgrowth in PC12 cells and activates p38 MAPK and ERK pathways
  • Inhibits harmaline tremor in a mouse model of essential tremor
  • Shows protective effects in the mouse model of PD induced by MPTP
  • Promotes branched-chain amino acid (BCAA) catabolism in rats