Learn More
Medchemexpress LLC Octanoic acid (Caprylic acid) | 124-07-2 | 99.9% | 144.21 | 100 G
SDP
Catalog No. 5000403706
Encompass_Preferred
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. Octanoic acid also acts as a tremor-suppressing agent and is intended for research use only.
  • Oily liquid with a slightly unpleasant rancid taste.
  • Used in the production of esters for perfumery.
  • Used in the manufacture of dyes.
  • Acts as a tremor-suppressing agent.
Catalog No. 50-004-03706 Supplier Medchemexpress LLC Supplier No. HY41417100G
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. Octanoic acid also acts as a tremor-suppressing agent and is intended for research use only.
  • Oily liquid with a slightly unpleasant rancid taste.
  • Used in the production of esters for perfumery.
  • Used in the manufacture of dyes.
  • Acts as a tremor-suppressing agent.