Learn More
Medchemexpress LLC Octanoic acid (Caprylic acid) | 124-07-2 | 99.9% | 144.21 | 250 G
SDP
Catalog No. 5000403221
Encompass_Preferred
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. It also acts as a tremor-suppressing agent, and is a human endogenous metabolite.
  • Used commercially in the production of esters for perfumery
  • Used in the manufacture of dyes
  • Acts as a tremor-suppressing agent
  • Purity of 99.85%
Catalog No. 50-004-03221 Supplier Medchemexpress LLC Supplier No. 250GHY41417
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is used commercially in the production of esters for perfumery and in the manufacture of dyes. It also acts as a tremor-suppressing agent, and is a human endogenous metabolite.
  • Used commercially in the production of esters for perfumery
  • Used in the manufacture of dyes
  • Acts as a tremor-suppressing agent
  • Purity of 99.85%