Learn More
Medchemexpress LLC Octanoic acid (Caprylic acid) | 124-07-2 | 99.9% | 144.21 | 5 G
SDP
Catalog No. 5000403624
Encompass_Preferred
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is commercially used in the production of esters for perfumery and in dye manufacturing. It also functions as a tremor-suppressing agent.
  • Oily liquid with a slightly unpleasant rancid taste.
  • Used in the production of esters for perfumery.
  • Utilized in dye manufacturing.
  • Functions as a tremor-suppressing agent.
Catalog No. 50-004-03624 Supplier Medchemexpress LLC Supplier No. HY414175G
Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste. It is commercially used in the production of esters for perfumery and in dye manufacturing. It also functions as a tremor-suppressing agent.
  • Oily liquid with a slightly unpleasant rancid taste.
  • Used in the production of esters for perfumery.
  • Utilized in dye manufacturing.
  • Functions as a tremor-suppressing agent.