Learn More
Apexbio Technology LLC OCTANOIC-ACID-D15-25MG
SDP
Catalog No. 5000570231
Encompass_Preferred
Octanoic acid-d15 (CAS No 69974-55-6) is a deuterated fatty acid commonly used in biomedical research for tracing metabolic pathways due to its stable isotope labeling It is derived from octanoic acid and has its mechanism of action linked to the modulation of lipid metabolism and mitochondrial function This compound typically demonstrates activity in the nanomolar to low micromolar range in various biochemical assays It is frequently applied in in vitro experiments involving cell lines to study cellular uptake fatty acid oxidation and energy production pathways Additionally its use extends to animal studies and drug screening applications especially in research focused on metabolic disorders neurodegenerative diseases and energy metabolism Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70231 Supplier Apexbio Technology LLC Supplier No. SL101925
Octanoic acid-d15 (CAS No 69974-55-6) is a deuterated fatty acid commonly used in biomedical research for tracing metabolic pathways due to its stable isotope labeling It is derived from octanoic acid and has its mechanism of action linked to the modulation of lipid metabolism and mitochondrial function This compound typically demonstrates activity in the nanomolar to low micromolar range in various biochemical assays It is frequently applied in in vitro experiments involving cell lines to study cellular uptake fatty acid oxidation and energy production pathways Additionally its use extends to animal studies and drug screening applications especially in research focused on metabolic disorders neurodegenerative diseases and energy metabolism Experimental concentrations typically depend on the specific research design