Learn More
Medchemexpress LLC Octanoic acid-d15 | 69974-55-6 | 99.9% | 159.30 | 25 MG
SDP
Catalog No. 5000402360
Encompass_Preferred
Octanoic acid-d15 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters used in perfumery and also in the manufacture of dyes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-02360 Supplier Medchemexpress LLC Supplier No. 25MGHY41417S
Octanoic acid-d15 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters used in perfumery and also in the manufacture of dyes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.