Learn More
Medchemexpress LLC Octanoic acid-d15 | 69974-55-6 | 99.9% | 159.30 | 5 MG
SDP
Catalog No. 5000404416
Encompass_Preferred
Octanoic acid-d15 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid used in the production of esters for perfumery and in the manufacture of dyes. This compound serves as a valuable tool in quantitative analysis.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-04416 Supplier Medchemexpress LLC Supplier No. HY41417S5MG
Octanoic acid-d15 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid used in the production of esters for perfumery and in the manufacture of dyes. This compound serves as a valuable tool in quantitative analysis.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs