Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoic acid-d15 | 69974-55-6 | MFCD00145029 | 99.9% | 10 MG
SDP

Catalog No. 5000297881
Encompass_Preferred

Octanoic acid-d15 is the deuterium-labeled analogue of octanoic (caprylic) acid, provided as a small-mass analytical standard for use in mass spectrometry, isotope tracing, and quantitative analyses.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic/purity: ~99.9% (as listed).
  • Molecular weight 159.30 g/mol.
  • CAS number 69974-55-6.
  • Molecular formula C8HD15O2.
  • Supplied in small milligram quantities, typically 10 MG packs.

Catalog No. 50-002-97881 Supplier Medchemexpress LLC Supplier No. HY41417S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octanoic acid-d15 is the deuterium-labeled analogue of octanoic (caprylic) acid, provided as a small-mass analytical standard for use in mass spectrometry, isotope tracing, and quantitative analyses.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic/purity: ~99.9% (as listed).
  • Molecular weight 159.30 g/mol.
  • CAS number 69974-55-6.
  • Molecular formula C8HD15O2.
  • Supplied in small milligram quantities, typically 10 MG packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.