Learn More
Medchemexpress LLC Octanoic acid-d3 | 156779-05-4 | 99.1% | 147.23 | 25 MG
SDP
Catalog No. 5000402199
Encompass_Preferred
Octanoic acid-d3 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and used commercially in the production of esters used in perfumery and also in the manufacture of dyes.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-02199 Supplier Medchemexpress LLC Supplier No. 25MGHY41417S1
Add to Cart
Add to Cart
Octanoic acid-d3 is the deuterium labeled Octanoic acid. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and used commercially in the production of esters used in perfumery and also in the manufacture of dyes.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.