Learn More
Medchemexpress LLC Octanoic acid-d3 | 156779-05-4 | 99.1% | 147.23 | 50 MG
SDP
Catalog No. 5000404514
Encompass_Preferred
Octanoic acid-d3 (Caprylic acid-d3) is the deuterium labeled Octanoic acid. Octanoic acid is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters used in perfumery and also in the manufacture of dyes. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04514 Supplier Medchemexpress LLC Supplier No. HY41417S150MG
Octanoic acid-d3 (Caprylic acid-d3) is the deuterium labeled Octanoic acid. Octanoic acid is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters used in perfumery and also in the manufacture of dyes. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.