Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoic acid-d3 | 156779-05-4 | MFCD00190494 | 99.1% | 147.23 g/mol | C8H13D3O2 | 10 MG
SDP

Catalog No. 5000298584
Encompass_Preferred

Octanoic acid-d3 is the deuterium-labeled (d3) analogue of octanoic (caprylic) acid provided as a high-purity isotopically labeled reagent for analytical and tracer applications. It is supplied as an oily liquid in small-quantity vials intended for use as internal standards in quantitative analyses such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (99.1% d3).
  • Molecular weight 147.23 g/mol and formula C8H13D3O2.
  • Supplied in small quantities suitable for analytical workflows.
  • Suitable for NMR, GC-MS, LC-MS, and tracer studies.

Catalog No. 50-002-98584 Supplier Medchemexpress LLC Supplier No. HY41417S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octanoic acid-d3 is the deuterium-labeled (d3) analogue of octanoic (caprylic) acid provided as a high-purity isotopically labeled reagent for analytical and tracer applications. It is supplied as an oily liquid in small-quantity vials intended for use as internal standards in quantitative analyses such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic enrichment (99.1% d3).
  • Molecular weight 147.23 g/mol and formula C8H13D3O2.
  • Supplied in small quantities suitable for analytical workflows.
  • Suitable for NMR, GC-MS, LC-MS, and tracer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.