Learn More
Medchemexpress LLC Octanoic acid (Standard) | 124-07-2 | 98.95% | 144.21 | 250 MG
SDP
Catalog No. 5000402927
Encompass_Preferred
Octanoic acid (Standard) is the analytical standard of Octanoic acid, intended for research and analytical applications. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes. This product is for research use only and not sold to patients. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Suitable for research and analytical applications
  • Used in the production of esters for perfumery
  • Used in the manufacture of dyes
  • Reference standard for qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-02927 Supplier Medchemexpress LLC Supplier No. 250MGHY41417R
Octanoic acid (Standard) is the analytical standard of Octanoic acid, intended for research and analytical applications. Octanoic acid (Caprylic acid) is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes. This product is for research use only and not sold to patients. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Suitable for research and analytical applications
  • Used in the production of esters for perfumery
  • Used in the manufacture of dyes
  • Reference standard for qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS