Learn More
Medchemexpress LLC Octanoic acid (standard) | 124-07-2 | 98.95% | 144.21 | 50 MG
SDP
Catalog No. 5000404863
Encompass_Preferred
Octanoic acid (Standard) is the analytical standard of Octanoic acid. It is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes. This compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Reference standard supplied assay
  • Suitable for research and analytical applications
  • Used in HPLC, GC, and MS experiments
  • Utilized in perfumery and dye manufacturing
Catalog No. 50-004-04863 Supplier Medchemexpress LLC Supplier No. HY41417R50MG
Octanoic acid (Standard) is the analytical standard of Octanoic acid. It is an oily liquid with a slightly unpleasant rancid taste and is used commercially in the production of esters for perfumery and in the manufacture of dyes. This compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Reference standard supplied assay
  • Suitable for research and analytical applications
  • Used in HPLC, GC, and MS experiments
  • Utilized in perfumery and dye manufacturing