Learn More
Octanoic-d15 Acid, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 69974-55-6 |
| Physical Form | Neat |
| Linear Formula | CD3(CD2)6 CO2H |
| MDL Number | MFCD00145029 |
| Melting Point | 16°C |
| Boiling Point | 237°C |
| Synonym | Caprylic-d15 acid |
| Quantity | 25 mg |
| Formula Weight | 159.30 |
| Percent Purity | ≥99.0% (GC) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.