Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octanoyl chloride

Catalog No. O47335ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O47335ML 5 mL
O4733100ML 100 mL
O4733500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O47335ML Supplier Sigma Aldrich Supplier No. O47335ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Applications of octanoyl chloride include:
  • Synthesis of (r)-2-propyloctanoic acid, a therapeutic agent for alzheimerαs disease
  • Synthesis of a variety of n-n-octyl-d-gluconamide based organogels
  • Building 2-heptylbenzo[d]thiazole moiety to synthesize conjugated copolymer with fluorine, as cathode interlayer in inverted polymer solar cells (pscs)
  • Octanoyl chloride is one of the essential precursors in the total synthesis of ()-mandelalide L, a marine macrolide that display significant cytotoxicity against human cancer cell lines

Specifications

CAS 111-64-8
Density 0.953 g/mL (at 25°C (literature))
Boiling Point 195°C (lit.)
Molecular Formula C8H15ClO
Linear Formula CH3(CH2)6 COCl
Refractive Index n20/D 1.435 (literature)
MDL Number MFCD00000766
Quantity 5 mL
Synonym Capryloyl chloride
Molecular Weight (g/mol) 162.66
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.