Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoyl coenzyme A lithium salt hydrate | 324518-20-9 | MFCD10567452 | 99.0% | 917.7 g/mol | C29H50N7O17P3S·xLi | 5 MG
SDP

Catalog No. 5000262620
Encompass_Preferred

Octanoyl coenzyme A lithium salt hydrate is the lithium salt form of octanoyl-coenzyme A, a medium-chain fatty acyl-coenzyme A used in biochemical and enzymology research. It serves as an acyl donor for peptide modification and is reported to inhibit citrate synthase and glutamate dehydrogenase, making it useful for studies of fatty-acyl-CoA metabolism and enzyme regulation.

  • Lithium salt hydrate of octanoyl-coenzyme A (CAS 324518-20-9).
  • Acts as an acyl donor for peptide octanoylation and enzymatic reactions.
  • Inhibits citrate synthase and glutamate dehydrogenase (reported IC50 0.4-1.6 mM).
  • Provided in small research quantities suitable for biochemical experiments.
  • Available as high-purity material (commonly ≥95%).
  • Chemical formula reported as C29H50N7O17P3S·xLi (hydrate).

Catalog No. 50-002-62620 Supplier Medchemexpress LLC Supplier No. HY134136A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Octanoyl coenzyme A lithium salt hydrate is the lithium salt form of octanoyl-coenzyme A, a medium-chain fatty acyl-coenzyme A used in biochemical and enzymology research. It serves as an acyl donor for peptide modification and is reported to inhibit citrate synthase and glutamate dehydrogenase, making it useful for studies of fatty-acyl-CoA metabolism and enzyme regulation.

  • Lithium salt hydrate of octanoyl-coenzyme A (CAS 324518-20-9).
  • Acts as an acyl donor for peptide octanoylation and enzymatic reactions.
  • Inhibits citrate synthase and glutamate dehydrogenase (reported IC50 0.4-1.6 mM).
  • Provided in small research quantities suitable for biochemical experiments.
  • Available as high-purity material (commonly ≥95%).
  • Chemical formula reported as C29H50N7O17P3S·xLi (hydrate).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.