Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octanoyl-L-carnitine | 25243-95-2 | MFCD28580111 | ≥99.0% | 287.40 g/mol | C15H29NO4 | 5 MG
SDP

Catalog No. 5000263750
Encompass_Preferred

L-Octanoylcarnitine is an endogenous human plasma metabolite and the physiologically active O-octanoyl ester of L-carnitine supplied for research use. It is provided as a white to off-white solid and is used in studies of fatty acid metabolism and cancer research, including breast cancer investigations.

  • CAS number 25243-95-2.
  • Molecular formula C15H29NO4; molecular weight 287.40 g/mol.
  • Purity ≥99.0%.
  • Soluble in DMSO at about 100 mg/mL with ultrasonic aid; example in vivo formulations yield ≥2.5 mg/mL.
  • Physical form white to off-white solid; hygroscopic DMSO affects solubility.
  • Intended for research applications in endogenous metabolism and cancer studies.

Catalog No. 50-002-63750 Supplier Medchemexpress LLC Supplier No. HY1131615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Octanoylcarnitine is an endogenous human plasma metabolite and the physiologically active O-octanoyl ester of L-carnitine supplied for research use. It is provided as a white to off-white solid and is used in studies of fatty acid metabolism and cancer research, including breast cancer investigations.

  • CAS number 25243-95-2.
  • Molecular formula C15H29NO4; molecular weight 287.40 g/mol.
  • Purity ≥99.0%.
  • Soluble in DMSO at about 100 mg/mL with ultrasonic aid; example in vivo formulations yield ≥2.5 mg/mL.
  • Physical form white to off-white solid; hygroscopic DMSO affects solubility.
  • Intended for research applications in endogenous metabolism and cancer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.