Learn More
Medchemexpress LLC Octanoyl-L-carnitine-d3 chloride | 1334532-24-9 | MFCD22419179 | 99.9% | 326.87 g/mol | C15H27D3ClNO4 | 5 MG
SDP
Catalog No. 5000223093
Encompass_Preferred
Octanoyl-L-carnitine-d3 chloride is the deuterium-labeled chloride salt of octanoyl-L-carnitine supplied as a solid stable-isotope standard for analytical and research use. It is intended as an internal standard for LC-MS/MS and GC-MS assays and for metabolic tracing studies of fatty acid and carnitine metabolism. The product is provided in small milligram packs suitable for reference standards and method development.
  • Deuterium-labeled internal standard for mass spectrometry assays.
  • Provided as the chloride salt for compatibility with aqueous and organic solvents.
  • High purity suitable for quantitative analysis.
  • Solid, white to off-white appearance for easy handling and storage.
  • Supplied in small milligram quantities for reference standard use.
  • Useful for metabolic tracing and quantification of acylcarnitines.
Catalog No. 50-002-23093 Supplier Medchemexpress LLC Supplier No. HY139392S5MG
Octanoyl-L-carnitine-d3 chloride is the deuterium-labeled chloride salt of octanoyl-L-carnitine supplied as a solid stable-isotope standard for analytical and research use. It is intended as an internal standard for LC-MS/MS and GC-MS assays and for metabolic tracing studies of fatty acid and carnitine metabolism. The product is provided in small milligram packs suitable for reference standards and method development.
  • Deuterium-labeled internal standard for mass spectrometry assays.
  • Provided as the chloride salt for compatibility with aqueous and organic solvents.
  • High purity suitable for quantitative analysis.
  • Solid, white to off-white appearance for easy handling and storage.
  • Supplied in small milligram quantities for reference standard use.
  • Useful for metabolic tracing and quantification of acylcarnitines.