Learn More
Medchemexpress LLC Octanoylcarnitine chloride | 14919-35-8 | 98.0% | 323.86 g/mol | C15H30ClNO4 | 10 MG
SDP
Catalog No. 5000211900
Encompass_Preferred
Octanoylcarnitine chloride is a medium-chain acylcarnitine salt used as a biochemical research reagent. It is a homolog of acetylcarnitine chloride and is applied in studies of lipid metabolism, mitochondrial fatty acid transport, and gastrointestinal drug absorption.
  • Used in research of lipid metabolism and mitochondrial fatty acid transport.
  • Can enhance drug absorption in the gastrointestinal tract.
  • Supplied as a solid research reagent with reported purity of 98.0%.
  • Identified by CAS number 14919-35-8 and molecular weight ~323.86 g/mol.
  • Suitable for biochemical assays and metabolic studies.
Catalog No. 50-002-11900 Supplier Medchemexpress LLC Supplier No. HY10101410MG
Octanoylcarnitine chloride is a medium-chain acylcarnitine salt used as a biochemical research reagent. It is a homolog of acetylcarnitine chloride and is applied in studies of lipid metabolism, mitochondrial fatty acid transport, and gastrointestinal drug absorption.
  • Used in research of lipid metabolism and mitochondrial fatty acid transport.
  • Can enhance drug absorption in the gastrointestinal tract.
  • Supplied as a solid research reagent with reported purity of 98.0%.
  • Identified by CAS number 14919-35-8 and molecular weight ~323.86 g/mol.
  • Suitable for biochemical assays and metabolic studies.