Learn More
Medchemexpress LLC Octanoylcarnitine chloride | 14919-35-8 | 98.0% | 323.86 g/mol | C15H30ClNO4 | 5 MG
SDP
Catalog No. 5000211910
Encompass_Preferred
Octanoylcarnitine chloride is the octanoyl ester of carnitine supplied as the chloride salt and intended for laboratory research use. It is a medium-chain acylcarnitine used as a metabolic probe to study fatty acid transport and acylcarnitine-related pathways, and may influence gastrointestinal drug absorption.
  • High purity (≈98.0%).
  • Molecular formula C15H30ClNO4; molecular weight 323.86 g/mol.
  • Solid form suitable for analytical and biochemical assays.
  • Used as a metabolic probe for medium-chain acylcarnitine pathways.
  • Store according to supplier SDS to maintain stability.
Catalog No. 50-002-11910 Supplier Medchemexpress LLC Supplier No. HY1010145MG
Octanoylcarnitine chloride is the octanoyl ester of carnitine supplied as the chloride salt and intended for laboratory research use. It is a medium-chain acylcarnitine used as a metabolic probe to study fatty acid transport and acylcarnitine-related pathways, and may influence gastrointestinal drug absorption.
  • High purity (≈98.0%).
  • Molecular formula C15H30ClNO4; molecular weight 323.86 g/mol.
  • Solid form suitable for analytical and biochemical assays.
  • Used as a metabolic probe for medium-chain acylcarnitine pathways.
  • Store according to supplier SDS to maintain stability.