Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octaphenyloctasilsesquioxane, TCI America™
SDP

Catalog No. O03495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03495G Supplier TCI America Supplier No. O03495G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5256-79-1
Molecular Formula C48H40O12Si8
Molecular Weight (g/mol) 1033.516
MDL Number MFCD00308870
InChI Key KBXJHRABGYYAFC-UHFFFAOYSA-N
Synonym 1,3,5,7,9,11,13,15-Octaphenylpentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane
PubChem CID 521391
SMILES C1=CC=C(C=C1)[Si]23O[Si]4(O[Si]5(O[Si](O2)(O[Si]6(O[Si](O3)(O[Si](O4)(O[Si](O5)(O6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 500°C
Color White
Quantity 5 g
Formula Weight 1033.52
Physical Form Crystalline Powder
Chemical Name or Material Octaphenyloctasilsesquioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.