Learn More
Medchemexpress LLC HY-B2170A 200mg Medchemexpress, Octenidine (dihydrochloride) CAS:70775-75-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2170A200MG
Encompass_Preferred
Medchemexpress, HY-B2170A 200mg Octenidine (dihydrochloride) CAS:70775-75-6 Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3093