Learn More
Selleck Chemical LLC Octenidine Dihydrochloride S5012-25mg
SDP
Supplier:  Selleck Chemical LLC S501225MG
Encompass_Preferred
Octenidine a cationic surfactant is active against gram-positive and gram-negative bacteria
Catalog No. 50-311-0933