Learn More
Apexbio Technology LLC Octenidine (dihydrochloride) 70775-75-6 1g
SDP
Catalog No. 5000092697
Encompass_Preferred
Octenidine dihydrochloride (CAS 70775-75-6) is a small-molecule inhibitor designed to disrupt microbial membrane integrity thereby inhibiting bacterial proliferation Octenidine dihydrochloride exerts its biological activity primarily through disruption of microbial membrane integrity In in vitro studies Octenidine dihydrochloride demonstrates inhibitory activity against diverse Gram-positive and Gram-negative bacterial strains with reported IC50 values typically ranging from 1 to 10 g/mL depending on test organisms and experimental conditions Based on these pharmacological properties Octenidine dihydrochloride holds research potential in infection control wound management and microbiological assay applications
Catalog No. 50-000-92697 Supplier Apexbio Technology LLC Supplier No. C64321000
Add to Cart
Add to Cart
Octenidine dihydrochloride (CAS 70775-75-6) is a small-molecule inhibitor designed to disrupt microbial membrane integrity thereby inhibiting bacterial proliferation Octenidine dihydrochloride exerts its biological activity primarily through disruption of microbial membrane integrity In in vitro studies Octenidine dihydrochloride demonstrates inhibitory activity against diverse Gram-positive and Gram-negative bacterial strains with reported IC50 values typically ranging from 1 to 10 g/mL depending on test organisms and experimental conditions Based on these pharmacological properties Octenidine dihydrochloride holds research potential in infection control wound management and microbiological assay applications