Learn More
Apexbio Technology LLC Octenidine (dihydrochloride) 70775-75-6 200mg
SDP
Catalog No. 5000089683
Encompass_Preferred
Octenidine dihydrochloride (CAS 70775-75-6) is a small-molecule inhibitor designed to disrupt microbial membrane integrity thereby inhibiting bacterial proliferation Octenidine dihydrochloride exerts its biological activity primarily through disruption of microbial membrane integrity In in vitro studies Octenidine dihydrochloride demonstrates inhibitory activity against diverse Gram-positive and Gram-negative bacterial strains with reported IC50 values typically ranging from 1 to 10 g/mL depending on test organisms and experimental conditions Based on these pharmacological properties Octenidine dihydrochloride holds research potential in infection control wound management and microbiological assay applications
Catalog No. 50-000-89683 Supplier Apexbio Technology LLC Supplier No. C6432200
Add to Cart
Add to Cart
Octenidine dihydrochloride (CAS 70775-75-6) is a small-molecule inhibitor designed to disrupt microbial membrane integrity thereby inhibiting bacterial proliferation Octenidine dihydrochloride exerts its biological activity primarily through disruption of microbial membrane integrity In in vitro studies Octenidine dihydrochloride demonstrates inhibitory activity against diverse Gram-positive and Gram-negative bacterial strains with reported IC50 values typically ranging from 1 to 10 g/mL depending on test organisms and experimental conditions Based on these pharmacological properties Octenidine dihydrochloride holds research potential in infection control wound management and microbiological assay applications