Learn More
Medchemexpress LLC Octenidine dihydrochloride | 70775-75-6 | 99.7% | 5 G
SDP
Catalog No. 5000412350
Encompass_Preferred
Octenidine dihydrochloride is an effective antiseptic compound for skin, mucous membranes, and wounds. It is for research use only and not sold to patients.
  • Effective antiseptic compound
  • Reduces resident microflora populations by 90 to 99.98%
  • More effective than chlorhexidine in reducing plaque score on buccal tooth surfaces
Catalog No. 50-004-12350 Supplier Medchemexpress LLC Supplier No. HYB2170A5G
Octenidine dihydrochloride is an effective antiseptic compound for skin, mucous membranes, and wounds. It is for research use only and not sold to patients.
  • Effective antiseptic compound
  • Reduces resident microflora populations by 90 to 99.98%
  • More effective than chlorhexidine in reducing plaque score on buccal tooth surfaces