Learn More
Cayman Chemical Octenidine (hydrochloride), Purity: ≥98%, CAS: 70775-75-6, 25 g
SDP
Catalog No. 502946424
Encompass_Preferred
An antimicrobial agent; active against methicillin-sensitive and -resistant S. aureus, vancomycin-resistant Enterococcus, E. faecalis, S. pneumoniae, and C. albicans (MICs = 1-32 mg/L); reduces methicillin- or vancomycin-resistant S. aureus biofilm formation at 2 mM; reduces dental plaque accumulation in cynomolgus monkeys fed a high-sucrose diet at 1%
Catalog No. 50-294-6424 Supplier Cayman Chemical Supplier No. 3423825 G
An antimicrobial agent; active against methicillin-sensitive and -resistant S. aureus, vancomycin-resistant Enterococcus, E. faecalis, S. pneumoniae, and C. albicans (MICs = 1-32 mg/L); reduces methicillin- or vancomycin-resistant S. aureus biofilm formation at 2 mM; reduces dental plaque accumulation in cynomolgus monkeys fed a high-sucrose diet at 1%