Learn More
Medchemexpress LLC Octenyl succinic anhydride | 26680-54-6 | 99.7% | C12H18O3 | 25 G
SDP
Catalog No. 5000352821
Encompass_Preferred
Octenyl succinic anhydride is a versatile chemical intermediate used in the manufacture of substances and as a laboratory chemical. It can undergo esterification with polysaccharides like starch and inulin, affecting intermolecular interactions on starch granule surfaces by altering their molecular structure.
  • Can be used for starch esterification
  • Modifies properties of polysaccharides
  • Produces OS-starch, an amphiphilic hydrocolloid
Catalog No. 50-003-52821 Supplier Medchemexpress LLC Supplier No. 25GHY145942
Octenyl succinic anhydride is a versatile chemical intermediate used in the manufacture of substances and as a laboratory chemical. It can undergo esterification with polysaccharides like starch and inulin, affecting intermolecular interactions on starch granule surfaces by altering their molecular structure.
  • Can be used for starch esterification
  • Modifies properties of polysaccharides
  • Produces OS-starch, an amphiphilic hydrocolloid