Learn More
U.S. Pharmacopeia Octinoxate, 5466-77-3, MFCD00072582, 500mg
Supplier:  U.S. Pharmacopeia 1477900
Encompass
Molecular formula C18H26O3, Molecular weight 290.4 g/mol, Melting Point 32 - -13 °F (0 - -25 °C), Boiling Point 590 °F (310 °C), Synonyms: Octyl methoxycinnamate
Catalog No. NC9175101