Learn More
Medchemexpress LLC Octisalate | 118-60-5 | 98.0% | 200 MG
SDP
Catalog No. 5000409387
Encompass_Preferred
Octisalate is an organic compound with high lipophilicity, primarily used in sunscreens and cosmetics for its ability to absorb UVB rays. This product is intended for research use only.
  • Used in sunscreens and cosmetics for absorbing UVB rays.
  • Features high lipophilicity.
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic.
  • Shows an average total absorption of skin permeation of 0.65% in vitro.
  • Stable under recommended storage conditions.
Catalog No. 50-004-09387 Supplier Medchemexpress LLC Supplier No. HYB0929200MG
Octisalate is an organic compound with high lipophilicity, primarily used in sunscreens and cosmetics for its ability to absorb UVB rays. This product is intended for research use only.
  • Used in sunscreens and cosmetics for absorbing UVB rays.
  • Features high lipophilicity.
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic.
  • Shows an average total absorption of skin permeation of 0.65% in vitro.
  • Stable under recommended storage conditions.