Learn More
U.S. Pharmacopeia Octisalate, 118-60-5, MFCD00053300, 400 mg
Supplier:  U.S. Pharmacopeia 1477943
Encompass
Linear Formula: (HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3, Molecular Weight: 250.33, bp: 189-190 °C/21 mmHg, Synonym: 2-Ethylhexyl salicylate, Octisalate, Octyl salicylate.
Catalog No. NC1282195